missing translation for 'onlineSavingsMsg'
Learn More

Hydrobromic acid, ACS, 47.0-49.0%

Catalog No. p-3208440
Change view
Click to view available options
Quantity:
250 mL
1 L
4 x 1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA36694AK 250 mL
AA36694K2 1 L
AA36694M1 4 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA36694AK Supplier Thermo Scientific Chemicals Supplier No. 036694AK
Only null left

Air Sensitive | CAS: 10035-10-6 | BrH | 80.91 g/mol

Hydrobromic acid is an indispensable raw material for organic intermediates, inorganic pharmaceuticals, photosensitive materials, dyes, and medicines. It is also used in the production of bromine compounds. It is used as an important agent in the industrial processes of isomerization, polymerization, hydration, and dehydration, and esterification reactions. It plays an essential role for the extraction of mercury from the cinnebar ore.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10035-10-6 , 7732-18-5
Molecular Formula BrH
Molecular Weight (g/mol) 80.91
MDL Number MFCD00011323
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-N
Synonym hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
PubChem CID 260
ChEBI CHEBI:47266
SMILES Br

Specifications

Appearance Colorless to pale yellow liquid
Color Colorless to Yellow
Density 1.49 g/cm3
Sensitivity Air Sensitive
Vapor Pressure 22500 hPa (16876mm Hg) at 20°C
Assay Percent Range 47.0-49.0%
Quantity 250 mL
Physical Form Liquid
Beilstein 3587158
Solubility Information Miscible in water.
IUPAC Name hydrogen bromide
Formula Weight 80.92
Grade ACS
Concentration or Composition (by Analyte or Components) Hydrogen bromide: 48.0%; Water: 52.0%
Chemical Name or Material Hydrobromic acid
Show More Show Less
Hazard Category H290-H314-H335
Precautionary Statement P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
DOTInformation Hazard Class: 8, Packing Group: II
EINECSNumber 233-113-0
RTECSNumber MW3850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only