missing translation for 'onlineSavingsMsg'
Learn More

Hydrobromic acid, 99.9999% (metals basis), 48% w/w aq. soln.

Catalog No. AA10991AP
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA10991AP 500 mL
AA10991AA 5 mL
AA10991AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA10991AP Supplier Thermo Scientific Chemicals Supplier No. 010991AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Air Sensitive | CAS: 10035-10-6 | BrH | 80.91 g/mol

Hydrobromic acid is used for the production of inorganic bromides of zinc, calcium, and sodium. It is used as a catalyst in some alkylation reactions. In the field of metallurgy, it is employed in the extraction of certain ores. Further, it is used as a reagent for the cleavage of certain ether linkages or as a brominating agent in organic transformations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10035-10-6 , 7732-18-5
Molecular Formula BrH
Molecular Weight (g/mol) 80.91
MDL Number MFCD00011323
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-N
Synonym hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
PubChem CID 260
ChEBI CHEBI:47266
SMILES Br

Specifications

Appearance Colorless to pale yellow liquid
Color Colorless to Yellow
Density 1.49
Sensitivity Air Sensitive
Vapor Pressure 22500 hPa (16876mm Hg)
Assay Percent Range 99.9999% (metals basis)
Quantity 500 mL
Beilstein 3587158
Physical Form Liquid
Solubility Information Miscible with water.
IUPAC Name bromane
Formula Weight 80.92
Concentration or Composition (by Analyte or Components) Hydrogen bromide: 48.0%; Water: 52.0%
Chemical Name or Material Hydrobromic acid
Show More Show Less
Hazard Category H290-H314-H335
Hazard Statement Keep container tightly closed and in a well-ventilated place. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
Precautionary Statement P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
DOTInformation Hazard Class: 8, Packing Group: II
EINECSNumber 233-113-0
RTECSNumber MW3850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only