missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrobromic Acid, 48%, Technical, Spectrum™ Chemical
Spécifications
Spécifications
| CAS Max % | 40 to 60% |
| Packaging | Poly-Coated Glass Bottle |
| Quantity | 500 mL |
| Grade | Technical |
| Chemical Name or Material | Hydrobromic Acid |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.