missing translation for 'onlineSavingsMsg'
Learn More

Hydrobromic Acid, 48%, Technical, Spectrum™ Chemical

Catalog No. 18610847
Change view
Click to view available options
Quantity:
500 mL
2.5 L
4 L
200 L
Packaging:
Poly-Coated Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610847 500 mL Poly-Coated Glass Bottle
18610844 2.5 L Poly-Coated Glass Bottle
18610846 4 L Poly-Coated Glass Bottle
18610845 200 L Poly-Coated Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18610847 Supplier Spectrum Chemical Mfg Cor Supplier No. H1031500MLGL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10035-10-6 , 7732-18-5

Specifications

CAS Max % 40 to 60%
Packaging Poly-Coated Glass Bottle
Quantity 500 mL
Grade Technical
Chemical Name or Material Hydrobromic Acid

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.