missing translation for 'onlineSavingsMsg'
Learn More

Hydrobromic Acid, 48%, Technical, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor H1031200LTBL

 View more versions of this product

Catalog No. 18610845

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

Hydrobromic Acid
40 to 60%,40 to 60%
200 L
10035-10-6,7732-18-5
Poly-Coated Glass Bottle
Technical

Chemical Identifiers

10035-10-6
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.