missing translation for 'onlineSavingsMsg'
Learn More

Hydrobromic Acid, 48%, ACS, 47.0-49.0%, Spectrum™ Chemical

Catalog No. 18610849
Change view
Click to view available options
Quantity:
120 mL
500 mL
2.5 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
Poly-Coated Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610849 500 mL Amber Glass Bottle
18610841 120 mL Amber Glass Bottle
18610848 2.5 L Poly-Coated Glass Bottle
18610840 4 L Amber Glass Bottle
18610843 20 L Poly Drum
18610842 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18610849 Supplier Spectrum Chemical Mfg Cor Supplier No. H1030500MLGL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10035-10-6 , 7732-18-5

Specifications

Concentration 48%
CAS Max % 40 to 60%
Heavy Metals 5 ppm
Packaging Amber Glass Bottle
Residue after Ignition 2e-05
Quantity 500 mL
Percent Purity 47 to 49%
Grade ACS
Chemical Name or Material Hydrobromic Acid

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.