missing translation for 'onlineSavingsMsg'
Learn More

Hydrobromic Acid, 48%, 47.0-49.0%, Spectrum™ Chemical

Catalog No. 18610836
Change view
Click to view available options
Quantity:
125 mL
500 mL
2.5 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
Poly-Coated Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610836 20 L Poly Drum
18610838 125 mL Amber Glass Bottle
18610839 500 mL Amber Glass Bottle
18610835 2.5 L Poly-Coated Glass Bottle
18610834 4 L Amber Glass Bottle
18610837 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18610836 Supplier Spectrum Chemical Mfg Cor Supplier No. H102920LTBL
Only null left

Chemical Identifiers

CAS 10035-10-6 , 7732-18-5

Specifications

CAS Max % 40 to 60%
Heavy Metals 5e-06
Packaging Poly Drum
Residue after Ignition 0.0002
Quantity 20 L
Percent Purity 47 to 49%
Grade Reagent
Chemical Name or Material Hydrobromic Acid

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.