missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrobromic Acid, 48%, 47.0-49.0%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 40 to 60% |
| Heavy Metals | 5e-06 |
| Packaging | Amber Glass Bottle |
| Residue after Ignition | 0.0002 |
| Quantity | 4 L |
| Percent Purity | 47 to 49% |
| Grade | Reagent |
| Chemical Name or Material | Hydrobromic Acid |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.