missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Hydrobenzoin 98.0+%, TCI America™

Catalog No. H08155G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08155G 5 g
H08151G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H08155G Supplier TCI America Supplier No. H08155G
Only null left

Chemical Identifiers

CAS 655-48-1
Molecular Formula C14H14O2
Molecular Weight (g/mol) 214.264
MDL Number MFCD00136059
InChI Key IHPDTPWNFBQHEB-UHFFFAOYSA-N
Synonym (+/-)-1,2-Diphenyl-1,2-ethanediol
PubChem CID 95447
ChEBI CHEBI:50013
IUPAC Name 1,2-diphenylethane-1,2-diol
SMILES C1=CC=C(C=C1)C(C(C2=CC=CC=C2)O)O

Specifications

Melting Point 123°C
Quantity 5 g
Formula Weight 214.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (+/-)-Hydrobenzoin