accessibility menu, dialog, popup

UserName

CAS RN 655-48-1

HOOH

CAS RN 655-48-1

IUPAC Name: 1,2-diphenylethane-1,2-diol
Synonyms: hydrobenzoin
Molecular Weight (g/mol): 214.26
Molecular Formula: C14H14O2
InChi Key: IHPDTPWNFBQHEB-UHFFFAOYNA-N
SMILES: OC(C(O)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results