Learn More
Micro Essential Lab Hydrion™ Double Roll pH Test Paper Dispensers
Hydrion™ Double Roll pH Papers consist of two rolls of pH papers packaged in a protective plastic dispenser with stainless steel cutoff lips and two matching color charts. pH color changes are distinct and easily matched to the color chart.
Supplier:  Micro Essential Lab 153
 View more versions of this product
Catalog No. 1485012D
Only null left
Description
Description
  • Features cut-off lips on each side
  • Wide-range papers provide a sharp color change for each full pH unit
  • Short-range papers change for each 0.5 pH unit
Each carton contains 10 dispensers, each consisting of two 15-foot rolls of test paper and matching color charts. Provides for approximately 2000 tests.
Directions
Simply tear off a small strip of pH paper, dip into the test solution, then instantly compare the resulting color with the matching pH color chart.
Specifications
Chemical Identifiers
Reusable dispenser, Matching color chart, 2 rolls of pH paper
4.6 m x 0.55 cm (15 ft. x 0.21 in.)
4.6 m
0.55 cm
2000 Tests
15 ft.
0.21 in.
Specifications
6 to 11
2000 Tests
4.6 m x 0.55 cm (15 ft. x 0.21 in.)
4.6 m
0.55 cm
Reusable dispenser, Matching color chart, 2 rolls of pH paper
Wide-range pH Test Paper
15 ft.
0.21 in.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
This product is not intended to diagnose, treat, cure or prevent any disease or condition. Please contact a medical doctor to diagnose and treat any medical condition.