Learn More
Micro Essential Lab Hydrion™ Double Roll pH Test Paper Dispensers
Hydrion™ Double Roll pH Papers consist of two rolls of pH papers packaged in a protective plastic dispenser with stainless steel cutoff lips and two matching color charts. pH color changes are distinct and easily matched to the color chart.
Supplier:  Micro Essential Lab MES150
 View more versions of this product
Catalog No. 1485012A
Only null left
Description
Description
  • Features cut-off lips on each side
  • Wide-range papers provide a sharp color change for each full pH unit
  • Short-range papers change for each 0.5 pH unit
Each carton contains 10 dispensers, each consisting of two 15-foot rolls of test paper and matching color charts. Provides for approximately 2000 tests.
Directions
Simply tear off a small strip of pH paper, dip into the test solution, then instantly compare the resulting color with the matching pH color chart.
Specifications
Chemical Identifiers
Reusable dispenser, Matching color chart, 2 rolls of pH paper
4.6 m x 0.55 cm (15 ft. x 0.21 in.)
4.6 m
0.55 cm
2000 Tests
15 ft.
0.21 in.
Specifications
1 to 11
2000 Tests
AB Wide-range pH Test Paper
15 ft.
0.21 in.
Reusable dispenser, Matching color chart, 2 rolls of pH paper
Roll No. 1 (Type A) pH range 2-4-6-8-10; Roll No. 2 (Type B) pH range 1-3-5-7-9-11
4.6 m x 0.55 cm (15 ft. x 0.21 in.)
4.6 m
0.55 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
This product is not intended to diagnose, treat, cure or prevent any disease or condition. Please contact a medical doctor to diagnose and treat any medical condition.