Learn More
Micro Essential Lab Hydrion™ Double Roll pH Test Paper Dispensers
Hydrion™ Double Roll pH Papers consist of two rolls of pH papers packaged in a protective plastic dispenser with stainless steel cutoff lips and two matching color charts. pH color changes are distinct and easily matched to the color chart.
Supplier: Micro Essential Lab MES8 12
Description
- Features cut-off lips on each side
- Wide-range papers provide a sharp color change for each full pH unit
- Short-range papers change for each 0.5 pH unit
Each carton contains 10 dispensers, each consisting of two 15-foot rolls of test paper and matching color charts. Provides for approximately 2000 tests.
Directions
Simply tear off a small strip of pH paper, dip into the test solution, then instantly compare the resulting color with the matching pH color chart.
Chemical Identifiers
| Reusable dispenser, Matching color chart, 2 rolls of pH paper | |
| 4.6 m x 0.55 cm (15 ft. x 0.21 in.) | |
| 4.6 m | |
| 0.55 cm |
| 2000 Tests | |
| 15 ft. | |
| 0.21 in. |
Specifications
| 8 to 12 | |
| 2000 Tests | |
| 4.6 m x 0.55 cm (15 ft. x 0.21 in.) | |
| 4.6 m | |
| 0.55 cm |
| Reusable dispenser, Matching color chart, 2 rolls of pH paper | |
| Wide-range pH Test Paper | |
| 15 ft. | |
| 0.21 in. |
This product is not intended to diagnose, treat, cure or prevent any disease or condition. Please contact a medical doctor to diagnose and treat any medical condition.