missing translation for 'onlineSavingsMsg'
Learn More

Micro Essential Lab Hydrion™ Buffer Chemvelopes

Catalog No. p-2537823
Change view
Click to view available options
pH:
10
11
12
2
3
4
6.4
7
8
9
Physical Form:
Capsule
Chemvelope
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Physical Form
14853258 12 Capsule
13640303H 10 Capsule
13640303J 11 Capsule
13640303A 2 Capsule
13640303B 3 Capsule
13640303C 4 Capsule
13640303E 7 Capsule
13640303F 8 Capsule
13640303G 9 Capsule
13640303D 6.4 Capsule
154202X - Capsule
154202W - Chemvelope
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 14853258 Supplier Micro Essential Lab Supplier No. 27012.00
Only null left

Powdered ingredients make precision buffers: ±0.02pH at 25°C

  • In capsules or packets (Chemvelopes)
  • Dissolve in distilled water: 100mL for capsules, 500mL for packets
  • Directly traceable to NIST standard

Standard Capsules come in vials of 10, five vials per pack. Tri-Chek Capsule Set includes one vial of 10 capsules each for pH 4, 7, and 10 buffers
Tri-Chek Chemvelope Set includes three pouches each for pH 4 and 10, four pouches for pH 7
Tri-Chek sets also include dispensing dropper bottle of Color Key/Buffer Preservative; add to buffer to inhibit mold and to a color-coded solution: red-orange for pH 4, green for pH 7, blue for pH 10

Specifications

pH 12
Physical Form Capsule
Quantity 5 Vials