Learn More
Hydrazine sulfate, ACS reagent
CAS: 10034-93-2 | H4N2·H2SO4 | 130.12 g/mol
Supplier: Thermo Scientific Chemicals 423771000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentSpecifications
| Hydrazine sulfate | |
| 10034-93-2 | |
| Crystalline Powder | |
| H4N2·H2SO4 | |
| MFCD00044873 | |
| hydrazine sulfate, hydrazine monosulfate, hydrazine, sulfate, hydrazine sulphate, hydrazinium sulfate, hydrazonium sulfate, hydrazine sulfate 1:1, segidrin, sehydrin, diamine sulfate | |
| ZGCHATBSUIJLRL-UHFFFAOYSA-N | |
| hydrazine;sulfuric acid | |
| 24842 | |
| ≥99.0% | |
| Plastic bottle |
| 254.0°C | |
| White | |
| 100 g | |
| H2NNH2·H2SO4 | |
| 15, 4810 | |
| Solubility in water: 30g/L (20°C). Other solubilities: insoluble in alcohol | |
| NN.OS(=O)(=O)O | |
| 130.12 | |
| 130.12 | |
| ACS Reagent | |
| 0.05% max |
Chemical Identifiers
| 10034-93-2 | |
| 130.12 | |
| ZGCHATBSUIJLRL-UHFFFAOYSA-N | |
| 24842 | |
| NN.OS(=O)(=O)O |
| H4N2·H2SO4 | |
| MFCD00044873 | |
| hydrazine sulfate, hydrazine monosulfate, hydrazine, sulfate, hydrazine sulphate, hydrazinium sulfate, hydrazonium sulfate, hydrazine sulfate 1:1, segidrin, sehydrin, diamine sulfate | |
| hydrazine;sulfuric acid |
Safety and Handling
GHS H Statement
May cause cancer.
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre
GHS Signal Word: Danger
EINECSNumber : 233-110-4
RUO – Research Use Only