missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine sulfate, ACS reagent

Catalog No. p-3756715
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC423770500 50 g Plastic bottle
AC423771000 100 g Plastic bottle
AC423775000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC423770500 Supplier Thermo Scientific Chemicals Supplier No. 423770500
Only null left

CAS: 10034-93-2 | H4N2·H2SO4 | 130.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent

Chemical Identifiers

CAS 10034-93-2
Molecular Formula H4N2·H2SO4
Molecular Weight (g/mol) 130.12
MDL Number MFCD00044873
InChI Key ZGCHATBSUIJLRL-UHFFFAOYSA-N
Synonym hydrazine sulfate, hydrazine monosulfate, hydrazine, sulfate, hydrazine sulphate, hydrazinium sulfate, hydrazonium sulfate, hydrazine sulfate 1:1, segidrin, sehydrin, diamine sulfate
PubChem CID 24842
IUPAC Name hydrazine;sulfuric acid
SMILES NN.OS(=O)(=O)O

Specifications

Melting Point 254.0°C
Color White
Physical Form Crystalline Powder
Quantity 50 g
Linear Formula H2NNH2·H2SO4
Merck Index 15, 4810
Solubility Information Solubility in water: 30g/L (20°C). Other solubilities: insoluble in alcohol
Formula Weight 130.12
Percent Purity ≥99.0%
Grade ACS Reagent
Packaging Plastic bottle
Residue after Ignition 0.05% max
Chemical Name or Material Hydrazine sulfate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause cancer.
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre
EINECSNumber 233-110-4

RUO – Research Use Only