missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrazine Monohydrochloride 98.0+%, TCI America™
$87.60 - $186.41
Chemical Identifiers
| CAS | 2644-70-4 |
|---|---|
| Molecular Formula | ClH5N2 |
| Molecular Weight (g/mol) | 68.504 |
| MDL Number | MFCD00044368 |
| InChI Key | BIVUUOPIAYRCAP-UHFFFAOYSA-N |
| Synonym | hydrazine hydrochloride, hydrazine monohydrochloride, hydrazine, monohydrochloride, hydrazine,chloride, diamine hydrochloride, hydrazine.hydrochloride, hydrazine,hydrochloride, hydrazine mono hcl, nh2nh2hcl, pubchem17422 |
| PubChem CID | 115031 |
| IUPAC Name | hydrazine;hydrochloride |
| SMILES | NN.Cl |
Chemical Identifiers
| 2644-70-4 | |
| 68.504 | |
| BIVUUOPIAYRCAP-UHFFFAOYSA-N | |
| 115031 | |
| NN.Cl |
| ClH5N2 | |
| MFCD00044368 | |
| hydrazine hydrochloride, hydrazine monohydrochloride, hydrazine, monohydrochloride, hydrazine,chloride, diamine hydrochloride, hydrazine.hydrochloride, hydrazine,hydrochloride, hydrazine mono hcl, nh2nh2hcl, pubchem17422 | |
| hydrazine;hydrochloride |
Specifications
| 93°C | |
| 2644-70-4 | |
| 25 g | |
| MFCD00044368 | |
| hydrazine hydrochloride, hydrazine monohydrochloride, hydrazine, monohydrochloride, hydrazine,chloride, diamine hydrochloride, hydrazine.hydrochloride, hydrazine,hydrochloride, hydrazine mono hcl, nh2nh2hcl, pubchem17422 | |
| NN.Cl | |
| 68.504 | |
| 68.50 | |
| Hydrazine Monohydrochloride |
| White | |
| Crystalline Powder | |
| ClH5N2 | |
| 3290 | |
| BIVUUOPIAYRCAP-UHFFFAOYSA-N | |
| hydrazine;hydrochloride | |
| 115031 | |
| ≥98.0% (T) |
Safety and Handling
EINECSNumber : (1)-0374
RTECSNumber : MV4625000
TSCA : Yes