missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrazine Monohydrochloride 98.0+%, TCI America™
Supplier: TCI America H0174500G
Specifications
| Hydrazine Monohydrochloride | |
| White | |
| Crystalline Powder | |
| ClH5N2 | |
| 3290 | |
| BIVUUOPIAYRCAP-UHFFFAOYSA-N | |
| hydrazine;hydrochloride | |
| 115031 | |
| ≥98.0% (T) |
| 93°C | |
| 2644-70-4 | |
| 500 g | |
| MFCD00044368 | |
| hydrazine hydrochloride, hydrazine monohydrochloride, hydrazine, monohydrochloride, hydrazine,chloride, diamine hydrochloride, hydrazine.hydrochloride, hydrazine,hydrochloride, hydrazine mono hcl, nh2nh2hcl, pubchem17422 | |
| NN.Cl | |
| 68.504 | |
| 68.50 |
Chemical Identifiers
| 2644-70-4 | |
| 68.504 | |
| BIVUUOPIAYRCAP-UHFFFAOYSA-N | |
| 115031 | |
| NN.Cl |
| ClH5N2 | |
| MFCD00044368 | |
| hydrazine hydrochloride, hydrazine monohydrochloride, hydrazine, monohydrochloride, hydrazine,chloride, diamine hydrochloride, hydrazine.hydrochloride, hydrazine,hydrochloride, hydrazine mono hcl, nh2nh2hcl, pubchem17422 | |
| hydrazine;hydrochloride |
Safety and Handling
EINECSNumber : (1)-0374
RTECSNumber : MV4625000
TSCA : Yes