missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrazine Dihydrochloride 98.0+%, TCI America™
$57.75 - $167.49
Chemical Identifiers
| CAS | 5341-61-7 |
|---|---|
| Molecular Formula | ClH5N2 |
| Molecular Weight (g/mol) | 68.50 |
| MDL Number | MFCD00064543 |
| InChI Key | BIVUUOPIAYRCAP-UHFFFAOYSA-N |
| Synonym | hydrazine dihydrochloride, hydrazine, dihydrochloride, unii-km8dd9j0c8, ccris 5479, diamine dihydrochloride, hydrazinium monochloride, km8dd9j0c8, hydrazine, hydrochloride 1:2, hydrazinedihydrochloride, hydrazine hcl |
| PubChem CID | 17548 |
| IUPAC Name | hydrogen hydrazine chloride |
| SMILES | [H+].[Cl-].NN |
Chemical Identifiers
| 5341-61-7 | |
| 68.50 | |
| BIVUUOPIAYRCAP-UHFFFAOYSA-N | |
| 17548 | |
| [H+].[Cl-].NN |
| ClH5N2 | |
| MFCD00064543 | |
| hydrazine dihydrochloride, hydrazine, dihydrochloride, unii-km8dd9j0c8, ccris 5479, diamine dihydrochloride, hydrazinium monochloride, km8dd9j0c8, hydrazine, hydrochloride 1:2, hydrazinedihydrochloride, hydrazine hcl | |
| hydrogen hydrazine chloride |
Specifications
| 5341-61-7 | |
| ClH5N2 | |
| 3290 | |
| 200°C | |
| BIVUUOPIAYRCAP-UHFFFAOYSA-N | |
| hydrogen hydrazine chloride | |
| 25 g | |
| ≥98.0% (T) |
| Powder/Crystals | |
| MFCD00064543 | |
| White | |
| hydrazine dihydrochloride, hydrazine, dihydrochloride, unii-km8dd9j0c8, ccris 5479, diamine dihydrochloride, hydrazinium monochloride, km8dd9j0c8, hydrazine, hydrochloride 1:2, hydrazinedihydrochloride, hydrazine hcl | |
| [H+].[Cl-].NN | |
| 68.50 | |
| 17548 | |
| Hydrazine Dihydrochloride |
Safety and Handling
EINECSNumber : (1)-0374
RTECSNumber : MV2298000
TSCA : Yes