missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine Dihydrochloride 98.0+%, TCI America™

Catalog No. H0170500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0170500G 500 g
H017025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0170500G Supplier TCI America Supplier No. H0170500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5341-61-7
Molecular Formula ClH5N2
Molecular Weight (g/mol) 68.50
MDL Number MFCD00064543
InChI Key BIVUUOPIAYRCAP-UHFFFAOYSA-N
Synonym hydrazine dihydrochloride, hydrazine, dihydrochloride, unii-km8dd9j0c8, ccris 5479, diamine dihydrochloride, hydrazinium monochloride, km8dd9j0c8, hydrazine, hydrochloride 1:2, hydrazinedihydrochloride, hydrazine hcl
PubChem CID 17548
IUPAC Name hydrogen hydrazine chloride
SMILES [H+].[Cl-].NN

Specifications

Physical Form Powder/Crystals
UN Number 3290
Color White
Melting Point 200°C
Quantity 500 g
Percent Purity ≥98.0% (T)
Chemical Name or Material Hydrazine Dihydrochloride
EINECSNumber (1)-0374
RTECSNumber MV2298000
TSCA Yes