missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine acetate, 97%

Catalog No. AC411750050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC411750050 5 g Glass bottle
AC411750250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC411750050 Supplier Thermo Scientific Chemicals Supplier No. 411750050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7335-65-1 | C2H8N2O2 | 92.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent

Chemical Identifiers

CAS 7335-65-1
Molecular Formula C2H8N2O2
Molecular Weight (g/mol) 92.10
MDL Number MFCD00013141
InChI Key YFHNDHXQDJQEEE-UHFFFAOYSA-N
Synonym hydrazine acetate, hydrazinium acetate, hydrazine, monoacetate, hydrazine monoacetate, acetic acid; hydrazine, hydrazineacetate, hydrazine, acetate 1:1, acetic acid; diamine, hydrazine mono-acetate, acmc-209bph
PubChem CID 165591
IUPAC Name acetic acid;hydrazine
SMILES NN.CC(O)=O

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula NH2NH2·CH3COOH
Quantity 5 g
Formula Weight 92.1
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material Hydrazine acetate, 99%
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Very toxic to aquatic life with long lasting effects.
Toxic if inhaled.
Toxic if swallowed.
Toxic in contact with skin.
May cause cancer.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF ON SKIN: Gently wash with plenty of soap and water
EINECSNumber 230-845-2
TSCA TSCA

RUO – Research Use Only