accessibility menu, dialog, popup

UserName

CAS RN 7335-65-1

IUPAC Name: acetic acid; hydrazine
Synonyms: acetic acid; hydrazine
Molecular Weight (g/mol): 92.1
Molecular Formula: C2H8N2O2
InChi Key: YFHNDHXQDJQEEE-UHFFFAOYSA-N
SMILES: NN.CC(O)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results