missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRANAL™ - Water Standard 0.1, Standard for Karl Fischer titration (water content 0.1 mg/g = 0.01%), verified against NIST SRM 2890 & NMIJ CRM 4222, Solstice
Specifications
Specifications
| Boiling Point | 136°C to 145°C |
| Density | 0.86 g/mL at 20 °C |
| CAS | 7732-18-5, 95-47-6 |
| Quantity | 1 Kit |
| CAS Max % | ≤100.0000% |
| Flash Point | 23 − 27°C |
| Packaging | Ampoule |
| UN Number | UN1307 |
| Formula Weight | 106.17 |
| Grade | Karl Fischer |
| Show More |
Safety and Handling
| ShelfLife | 720 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 3; Packing Group:3: III; Proper Shipping Name: Xylenes |
| Recommended Storage | 2° to 8°C |