missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRANAL™ - Titrant 5, Reagent for volumetric two-component Karl Fischer titration, Solstice
Specifications
Specifications
| Chemical Name or Material | HYDRANAL™-Titrant 5 |
| Density | 0.85g/mL |
| Quantity | 500 mL |
| Flash Point | 11°C (51.8°F) |
| UN Number | UN1230 |