missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRANAL™ - Standard sodium tartrate dihydrate, Standard for Karl Fischer titration (water content ∼15.66%), Honeywell™ Fluka™
Description
Advantages of HYDRANAL-Water Standards:
- Broad product range for volumetric and coulometric Karl Fischer applications, as well as the indirect oven application
- Manufactured according to ISO 9001 requirements
- Report of Analysis showing the exact water content and expanded uncertainty
Verified against external standards such as NIST SRM 2890 or NMIJ CRM 4222
- Long shelf life (up to five years)
- Convenient packaging
- Supplied with detailed instruction for use
Specifications
Specifications
| CAS | 6106-24-7 |
| Quantity | 25 g |
| UN Number | NONH for all modes of transport |
| Chemical Name or Material | HYDRANAL™-Standard sodium tartrate dihydrate |