missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™-Standard 5.0, Honeywell Fluka™

Catalog No. 60046463
Change view
Click to view available options
Quantity:
500 mL
6 x 100 mL
6 x 500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60046463 6 x 500 mL
6000675 500 mL
60046462 6 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60046463 Supplier Honeywell Chemicals Supplier No. 348136X500ML
Only null left

Test solution for rough check of volumetric KF equipment.

Advantages of HYDRANAL- Auxiliaries

  • Broad product range of solubilizer for special sample application
  • Broad product range of buffer for pH related sample application
  • Drying agents for cell protection and drying of carrier gasses.

Specifications

Chemical Name or Material HYDRANAL™-Standard 5.0
CAS 71-36-3, 95-47-6
Quantity 6 x 500 mL
Flash Point 27°C (80.6°F)
UN Number UN1993C