Learn More
HYDRANAL™-Sodium tartrate dihydrate, Honeywell Fluka™
Test substance for volumetric Karl Fischer titration (water content ∼15.66%, exact value on report of analysis)
Supplier: Honeywell Chemicals 348036X100G
Description
Advantages of HYDRANAL- Auxiliaries
- Broad product range of solubilizer for special sample application
- Broad product range of buffer for pH related sample application
- Drying agents for cell protection and drying of carrier gasses.
Specifications
| HYDRANAL™-Sodium tartrate dihydrate | |
| 6106-24-7 | |
| 6 x 100 g | |
| NONH for all modes of transport |