missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™-Molecular sieve 0.3 nm, Solstice

Catalog No. 6002483
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002483 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6002483 Supplier Solstice Supplier No. 34241250G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Drying agent for air and gases for Karl Fischer titration

Advantages of HYDRANAL- Auxiliaries

  • Broad product range of solubilizer for special sample application
  • Broad product range of buffer for pH related sample application
  • Drying agents for cell protection and drying of carrier gasses.

Specifications

Chemical Name or Material HYDRANAL™-Molecular sieve 0.3 nm
Density >2 g/3
Pore Size 0.3 nm
Quantity 250 g
CAS 1318-02-1
UN Number NONH for all modes of transport