Learn More
HYDRANAL™ - Methanol dry, Reagent for volumetric one-component KF titration (working medium), Honeywell Fluka™
Supplier:  Honeywell Chemicals 347416X1LUS
 View more versions of this product
Catalog No. 6000659
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
HYDRANAL™-Methanol dry
67-56-1
9°C (48.2°F)
0.79g/mL
1 L
UN1230
Product Suggestions
Product Suggestions