Learn More
HYDRANAL™ - Medium K, Honeywell Fluka™
Medium for volumetric one-component Karl Fischer titration in aldehydes and ketones (methanol free).
$244.73 - $296.30
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
60-090-317
|
Honeywell Inc
346981LUS |
1 L |
Each for $296.30
|
|
|||||
|
6000658
|
Honeywell-Hydranal
346986X1LUS |
6 x 1 L |
Each for $244.73
Case of 6 Each for $1,468.38
|
|
|||||
Description
Advantages of Hydranal one-component reagents:
- Unlimited water capacity
- Convenient and easy to use
- The greatest flexibility in working media selection
- reduced toxicity compared to methanol based systems
- Suitable for methanol-reacting compounds, e.g. ketones and aldehydes
- Long shelf life (three years)
Specifications
| UN1992 |