Learn More
HYDRANAL™ - LipoSolver CM, Reagent for volumetric one-component KF titration in non-polar substances, fats and oils (working medium), Honeywell Fluka™
Medium for volumetric one-component Karl Fischer titration in non-polar substances, fats and oils (chloroform-methanol-based)
Supplier: Honeywell-Fluka 378556X1L
Description
Advantages of Hydranal one-component reagents:
- Unlimited water capacity
- Convenient and easy to use
- The greatest flexibility in working media selection
- reduced toxicity compared to methanol based systems
- Suitable for methanol-reacting compounds, e.g. ketones and aldehydes
- Long shelf life (three years)
Improved Composition
Hydranal-Composite contains all the reactants including iodine, sulfur dioxide, and the bases imidazole and 2-methylimidazole, dissolved in diethylene glycol monoethyl ether (DEGEE). Adding 2-methylimidazole in addition to imidazole improves the stability and eliminates the formation of crystals which can interfere with the titrator’s performance.
Specifications
| HYDRANAL™ - LipoSolver CM | |
| 54°C | |
| ∼1.25Â g/mL | |
| 6 x 1 L | |
| 26°C | |
| Glass Bottle | |
| Methylidyne trichloride |
| Reagent for volumetric one-component KF titration in non-polar substances, fats and oils (working medium) | |
| ≥70.0000%,≥20.0000% | |
| 67-56-1 | |
| <90.0000% | |
| MFCD02687786 | |
| UN1992 | |
| Karl Fischer |