missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRANAL™ - CRM Water Standard 1.0, Certified reference material for Karl Fischer titration (water content 1 mg/g = 0.1%), Solstice
Specifications
Specifications
| Boiling Point | 153°C to 155°C |
| CAS Min % | ≥90.0000%, ≥1.0000% |
| Density | 1.000 g/cm3 |
| CAS | 108-32-7, 100-66-3 |
| Quantity | 10 x 4 mL |
| CAS Max % | <100.0000% |
| Flash Point | 43°C |
| Packaging | Ampoule |
| UN Number | UN2222 |
| Grade | Certified Reference Material |
| Show More |
Safety and Handling
| ShelfLife | 1080 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 3; Packing Group:3: III; Proper Shipping Name: Anisole solution |