missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRANAL™ - Coulomat E, Reagent for coulometric KF titration (ethanol-based), for cells with and without diaphragm, Honeywell Fluka™
Description
Advantages of Hydranal Coulometric Reagents
- Easy to use
- High accuracy for trace amounts of water
- Stable conditions of the titration vessel
- Broad product range
- Long shelf life (up to five years)
Specifications
Specifications
| Chemical Name or Material | HYDRANAL™-Coulomat E |
| CAS | 67-56-1, 111-42-2, 64-17-5 |
| Quantity | 500 mL |
| Flash Point | 14°C (57.2°F) |
| Packaging | Glass bottle |
| UN Number | UN1993BE |