missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Coulomat E, Reagent for coulometric KF titration (ethanol-based), for cells with and without diaphragm, Honeywell Fluka™

Catalog No. 6000650
Change view
Click to view available options
Quantity:
500 mL
6 x 500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000650 500 mL
60046399 6 x 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6000650 Supplier Honeywell Chemicals Supplier No. 34726500ML
Only null left

Anolyte for coulometric Karl Fischer titration (ethanol-based), for cells with and without diaphragm

Advantages of Hydranal Coulometric Reagents

  • Easy to use
  • High accuracy for trace amounts of water
  • Stable conditions of the titration vessel
  • Broad product range
  • Long shelf life (up to five years)

Specifications

Chemical Name or Material HYDRANAL™-Coulomat E
CAS 67-56-1, 111-42-2, 64-17-5
Quantity 500 mL
Flash Point 14°C (57.2°F)
Packaging Glass bottle
UN Number UN1993BE