missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Coulomat AG-Oven, Reagent for coulometric KF titration with oven (anolyte solution), for cells with and without diaphragm, Honeywell Fluka

Catalog No. p-7111847
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000647 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6000647 Supplier Honeywell Chemicals Supplier No. 347396X500MLUS
Only null left

Specifications

Chemical Name or Material HYDRANAL™ - Coulomat AG-Oven
Name Note Reagent for coulometric KF titration with oven (anolyte solution), for cells with and without diaphragm
Boiling Point 64°C
CAS Min % ≥5.0000%, ≥20.0000%
CAS Max % <20.0000%, <10.0000%, <10.0000%, <30.0000%
CAS 111-42-2, 7446-09-5, 57-55-6, 288-32-4
Density 0.980 g/cm3
Quantity 500 mL
Flash Point 13°C
Packaging Glass Bottle
UN Number UN1230
Grade Karl Fischer
Show More Show Less