missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Coulomat AG, Reagent for coulometric KF titration (anolyte solution), for cells with and without diaphragm, Solstice

Catalog No. p-7072141
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000645 500 mL
6000644 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6000645 Supplier Solstice Supplier No. 348366X500MLUS
Only null left

Specifications

Chemical Name or Material HYDRANAL™-Coulomat AG
CAS 7446-09-5, 68007-08-9, 111-42-2, 67-56-1, 288-32-4
Quantity 500 mL
Flash Point 14°C (57.2°F)
UN Number UN1230