missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Coulomat AD, Reagent for coulometric KF titration (anolyte solution), for cells without diaphragm, Honeywell Fluka™

Catalog No. p-6980056
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000642 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6000642 Supplier Honeywell Chemicals Supplier No. 348106X500MLUS
Only null left

Specifications

Chemical Name or Material HYDRANAL™-Coulomat AD
CAS 7446-09-5, 68007-08-9, 288-32-4
Quantity 500 mL
Flash Point 11°C (51.8°F)
UN Number UN1230