missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™-Composite 5, Titrating agent for volumetric one-component Karl Fischer titration (methanol free), Solstice

Catalog No. 60090316
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60090316 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60090316 Supplier Honeywell Inc Supplier No. 348051LUS
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Titrating agent designed for volumetric one-component Karl Fischer titration.

Advantages of Hydranal one-component reagents:

  • Unlimited water capacity
  • Convenient and easy to use
  • The greatest flexibility in working media selection
  • reduced toxicity compared to methanol based systems
  • Suitable for methanol-reacting compounds, e.g. ketones and aldehydes
  • Long shelf life (three years)

Improved Composition

Hydranal-Composite contains all the reactants including iodine, sulfur dioxide, and the bases imidazole and 2-methylimidazole, dissolved in diethylene glycol monoethyl ether (DEGEE). Adding 2-methylimidazole in addition to imidazole improves the stability and eliminates the formation of crystals which can interfere with the titrator's performance.

Chemical Identifiers

Molecular Formula C4H6N2
Molecular Weight (g/mol) 82.11
MDL Number MFCD00005190
InChI Key LXBGSDVWAMZHDD-UHFFFAOYSA-N
IUPAC Name 2-methyl-1H-imidazole
SMILES CC1=NC=CN1

Specifications

Boiling Point 194°C (1.013 hPa)
CAS Min % ≥5.00, ≥50.00, ≥5.00
CAS Max % <10.00, <70.00, <10.00, <10.00
CAS 7446-09-5, 7553-56-2, 68007-08-9, 693-98-1, 288-32-4
Density 1.17 g/cm3 (25°C)
Physical Form Liquid
Quantity 1 L
Flash Point 99°C
Packaging Polycoated Bottle
Grade One Component Volumetry
Hazard Category Danger
Hazard Statement H351+H360+H372
Precautionary Statement P201+P202+P260+P264+P270+P281
Recommended Storage Room Temperature