missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Composite 5 K, Reagent for volumetric one-component Karl Fischer titration in aldehydes and ketones (methanol free), Solstice

Catalog No. 6002571
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
6 x 500 mL
6 x 1 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002571 500 mL
6002569 1 L
6002570 2.5 L
60046469 6 x 500 mL
60046468 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 6002571 Supplier Honeywell Inc Supplier No. 34816500MLUS
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 111-90-0

Specifications

Chemical Name or Material HYDRANAL™-Composite 5 K
CAS 288-32-4, 7446-09-5, 7553-56-2, 111-90-0, 68007-08-9
Packaging Poly-coated Bottle
Density 1.15g/mL
Quantity 500 mL
Flash Point 90°C (194°F)
Grade Karl Fischer