missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Composite 5, Reagent for volumetric one-component Karl Fischer titration (methanol free), Solstice

Catalog No. 6002565
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
500 mL
1 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002565 500 mL
6002563 1 L
6002564 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Catalog No. 6002565 missing translation for 'mfr' Honeywell Inc missing translation for 'supplierNo' 348056X500MLUS

Specifications

Chemical Name or Material HYDRANAL™-Composite 5
CAS 288-32-4, 68007-08-9, 7553-56-2, 111-90-0, 693-98-1
Density 1.15g/mL
Quantity 500 mL
Flash Point 90°C (194°F)