missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Composite 2, Reagent for volumetric one-component Karl Fischer titration (methanol free), Solstice

Catalog No. 6002568
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002568 500 mL
6002566 1 L
6002567 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6002568 Supplier Honeywell Inc Supplier No. 348066X500MLUS
Edge

Specifications

Chemical Name or Material HYDRANAL™-Composite 2
CAS 7446-09-5, 693-98-1, 288-32-4, 111-90-0, 68007-08-9
Density 1.11g/mL
Quantity 500 mL
Flash Point 90°C (194°F)