missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HYD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589459
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589459 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589459 Supplier Invitrogen™ Supplier No. PA589459

Rabbit Polyclonal Antibody

Immunogen sequence: SISAGIPKVG VLMESVWNMN DSCRFQLRSP ESLKNMEKAS KTTEAKPESK QEPVKTEMGP PPSPASTCSD ASSIASSASM PYKRRRSTPA PKEEEKVNEE QWSLREVVFV EDVKNVPVGK VLKVDGAYVA VKFPGTSSNT NCQNSSGPDA DPSSLLQDCR LLRIDELQVV KTGGTPKVPD CFQRTPKKLC IPEKTEILAV NVDSKGVHAV L; Positive Samples: U-87MG, 293T, Jurkat, HeLa; Cellular Location: Nucleus.

EDD (for E3 identified by Differential Display) is a progestin-regulated gene that was isolated from T-47D human breast cancer cells. Based on sequence homology, EDD appears to be a human homolog of the Drosophila hyperplastic discs (hyd) gene, a tumor suppressor gene that is required for control of imaginal disc growth. EDD contains a HECT domain in the carboxy terminus. HECT domain-containing proteins function as ubiquitin-protein ligases, or E3 enzymes. EDD has been shown to bind to ubiquitin, and like other HECT family proteins, may function as an E3 ubiquitin-protein ligase.

Specifications

Antigen HYD
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.92 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene UBR5
Gene Accession No. O95071
Gene Alias CH D1S3362; DD5; E3 identified by differential display; E3 ubiquitin-protein ligase UBR5; E3 ubiquitin-protein ligase, HECT domain-containing 1; ECGF1; Edd; Edd1; EDG1; EDG-1; extraembryonic development; HECT-type E3 ubiquitin transferase UBR5; hHYD; HYD; Hyperplastic discs protein homolog; KIAA0896; Progestin-induced protein; Rat100; S1P receptor 1; S1P receptor Edg-1; S1P1; S1PR1; ubiquitin protein ligase E3 component n-recognin 5; Ubr5
Gene Symbols UBR5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 550-760 of human UBR5 (NP_0569862).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51366
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less