missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Water for Injection (WFI) Quality Water

Numéro de catalogue. SH3022128
Change view
Click to view available options
Quantity:
10 L
20 L
50 L
100 L
200 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
SH3022128 200 L
SH3022124 10 L
SH3022125 20 L
SH3022126 50 L
SH3022127 100 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Numéro de catalogue. SH3022128 Fournisseur Cytiva Code fournisseur SH30221.28
Il en reste null

Supports cell culture growth and biomanufacturing process.

  • Format: Liquid

Identifiants chimiques

CAS 7732-18-5
Molecular Formula H2O
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
SMILES O

Spécifications

IUPAC Name water
Molecular Weight (g/mol) 18.02
Quantity 200 L
Chemical Name or Material Water
missing translation for 'shelfLife' 36 Months