missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ US Bovine Calf Serum

Catalog No. SH3007303HI
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3007303HI 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3007303HI Supplier Cytiva Supplier No. SH30073.03HI
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

A cost effective alternative to Fetal Bovine Serum (FBS). Cytiva HyClone™ US Bovine Calf Serum is defined, irradiated, heat inactivated serum of US origin that is filtered through three sequential 0.1μm pore size-rated filters.

HyClone Bovine Calf Serum, U.S. origin is collected by venipuncture from formula-fed veal calves, which produce exceptionally high levels of transferrin. Before dispensing, each lot of serum is pooled using true pool technology to ensure uniformity and consistency between bottles.

  • Sourced in the USA with complete traceability back to original source
  • Cost-effective alternative to FBS
  • Age of animal at time of collection approximately 16-24 weeks
  • Low in antibodies and high in growth factors
  • Virus panel testing according to 9 CFR 113.53

Specifications

Additional Assays Insulin Gamma Globulin Alkaline Phosphatase Lactate Dehydrogenase SGPT SGOT pH Albumin Blood Urea Nitrogen Creatinine Total Bilirubin Sodium Potassium Calcium Chloride Inorganic Phosphorus Iron TIBC Percent Saturation Glucose IgG
Certifications/Compliance Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
Description Heat Inactivated; 500 mL
Species Bovine
Age 16 to 24 weeks
Country of Origin United States
Product Type Calf Bovine Serum
Quantity 500 mL
Sterility Sterile
Sterilization Method Sequential-filtered, 100 nm
Tested For Bluetongue Bovine Adenovirus Bovine Parvovirus Bovine RSV BVDV Cytopathogenic Agents Hemadsorbing Agents Rabies Virus Reovirus
Heat Inactivated Yes
USDA Safety Testing Included Yes
Show More Show Less