missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Trypan Blue Solution, 0.4% Phosphate Buffered Saline

Catalog No. SV3008401
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SV3008401 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SV3008401 Supplier Cytiva Supplier No. SV30084.01

Trypan Blue solution, 0,4%, is routinely used as a cell stain to assess cell viability using the dye exclusion test.

Trypan Blue solution, 0.4%, is routinely used as a cell stain to assess cell viability using the dye exclusion test. This test is often performed while counting cells with the hemocytometer during routine subculturing, but can be performed any time cell viability needs to be determined quickly and accurately. The dye exclusion test is based upon the concept that viable cells do not take up impermeable dyes (like Trypan Blue), but dead cells are permeable and take up the dye.

Specifications

Content And Storage 15° to 30°C
Form Liquid
Product Type Trypan Blue
Quantity 100 mL