missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Tetracycline Screened Characterized Fetal Bovine Serum, US Origin

Catalog No. SH3007103T
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3007103T 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3007103T Supplier Cytiva Supplier No. SH3007103T
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

FBS with undetectable levels of tetracycline

Tetracycline Screened FBS is designed for researchers using tetracycline-regulated gene expression systems in cultured cells and is suitable for expression studies in Tet-on/Tet-off systems.

Specifications

Country of Origin United States
Form Liquid
Quantity 500 mL
Processing None
Endotoxin Concentration ≤ 10 EU/mL
Heat Inactivated No
Hemoglobin Concentration ≤ 10 mg/dL