missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Tetracycline Screened Characterized Fetal Bovine Serum, US Origin
Description
Tetracycline Screened FBS is designed for researchers using tetracycline-regulated gene expression systems in cultured cells and is suitable for expression studies in Tet-on/Tet-off systems.
Specifications
Specifications
| Country of Origin | United States |
| Form | Liquid |
| Specialty Type | Tetracycline screened |
| Quantity | 500 mL |
| Processing | Heat Inactivated |
| Endotoxin Concentration | ≤ 10 EU/mL |
| Heat Inactivated | Yes |
| Hemoglobin Concentration | ≤ 10 mg/dL |