missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Tetracycline Screened Characterized Fetal Bovine Serum, US Origin

Catalog No. SH371TIH254
Change view
Click to view available options
Processing:
Heat Inactivated
HI, Irr - 25 to 40 doses
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Processing
SH371TIH254 HI, Irr - 25 to 40 doses
SH3007103TH Heat Inactivated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SH371TIH254 Supplier Cytiva Supplier No. SH30071.03TIH2540

FBS with undetectable levels of tetracycline

Tetracycline Screened FBS is designed for researchers using tetracycline-regulated gene expression systems in cultured cells and is suitable for expression studies in Tet-on/Tet-off systems.

Specifications

Country of Origin United States
Form Liquid
Quantity 500 mL
Processing HI, Irr - 25 to 40 doses
Endotoxin Concentration ≤ 10 EU/mL
Heat Inactivated Yes
Hemoglobin Concentration ≤ 10 mg/dL