missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ SG 200 solution

Catalog No. SH3059001
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3059001 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3059001 Supplier Cytiva Supplier No. SH30590.01
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Stable dipeptide of L-Alanyl L-Glutamine which acts as nutrient media supplement.

  • Used as nutrient media supplement

Specifications

Certifications/Compliance cGMP and ISO 91:2 Guidelines
Content And Storage 2° to 8°C
Description SG-200, (Stable dipeptide of L-AlanyL-Glutamine) 200mM, in 0.85% NaCl Solution
Concentration 2mM
Sterilization Method 0.1 μm Sterile-filtered
Endotoxin Level =1.0EU/mL
Quantity 100 mL