missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ SFM4Transfx-293 Powdered Media

Catalog No. SH3086101
Change view
Click to view available options
Quantity:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3086101 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3086101 Supplier Cytiva Supplier No. SH30861.01
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Promotes transfection using lipofection, polymer, or similar methods in HEK 293 cell culture.

HyClone SFMTransfx-293 medium is a serum-free and animal-derived component-free medium designed to support the growth of HEK 293 cell cultures and promote transfection using lipofection, polymer, or similar methods. This regulatory-friendly medium was developed to support high transfection efficiency, cell density, and productivity in suspension cultures.

SFM4Transfx-293 medium is available in liquid and powder formats.

  • Supports high transfection efficiency.
  • Allows for direct or sequential adaptation.
  • Animal-derived component-free.
  • Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities.
  • Designed for high cell yield and recombinant protein/vector production.

Specifications

Content And Storage 2 to 8°C, away from light and moisture
Form Powder
Sterility Non-sterile
Without Additives No Glutamine, No HEPES, No Phenol Red, No Sodium Bicarbonate
Quantity 5 L